Encapsulated chemcial protective suit for PAPR systems, AlphaTec® 3000 AIRline, Model 754
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
YE30T-00754-03 YE30T-00754-04 YE30T-00754-02 YE30T-00754-05 YE30T-00754-06 YE30T-00754-07
ANSEYE30T-00754-03EA 726 EUR
ANSEYE30T-00754-03 ANSEYE30T-00754-04 ANSEYE30T-00754-02 ANSEYE30T-00754-05 ANSEYE30T-00754-06 ANSEYE30T-00754-07
Encapsulated chemcial protective suit for PAPR systems, AlphaTec® 3000 AIRline, Model 754
Werkkleding
Encapsulated chemical protective suits for use with air-fed/supplied systems.
  • Exceptional protection; over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams offer the highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • HEPA filter provides secondary protection from airline contamination
  • MC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow)
  • Suit/belt mounted regulator complete with a low flow warning whistle
  • AVANT STS regulator can be mounted on the suit and removed for reuse
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached sock boots with anti-slip PVC sole allow soft or protective safety shoes to be worn inside the suit
Multi-layer non woven barrier laminate fabric with ultrasonically welded and taped seams. These fully encapsulated suits are designed for use in combination with the MICROCHEM AVANT STS V.1 airline regulator. Suit features include internal air-channel ventilation system double cuffs with the options of finger and thumb loop, G01 Ansell Solvex (Nitrile) gloves, G02 Ansell Barrier gloves, or G03 KCL Camapren (Neoprene) gloves. Attached boot ends with anti-slip PVC sole version also available. Emergency rip cord is standard.
Certificaten: CE Category III (Type 3, EN 1073-1, EN 14594, EN 1149-5)