HyFlex® 11-754 Cut-Resistant Industrial Gloves with INTERCEPT™ Technology
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11-754/10 11-754/11 11-754/12 11-754/6 11-754/7 11-754/8 11-754/9
ANSE11-754/10EA 264 EUR
ANSE11-754/10 ANSE11-754/11 ANSE11-754/12 ANSE11-754/6 ANSE11-754/7 ANSE11-754/8 ANSE11-754/9
HyFlex® 11-754 Cut-Resistant Industrial Gloves with INTERCEPT™ Technology
Handschoenen
Ansell's lightest ANSI A4/ISO Level D cut resistant touchscreen glove.
  • Touchscreen compatible to ensure that workers do not need to change or remove gloves between tasks
  • Designed to fit the natural shape of the hand to help reduce hand fatigue
  • Clean and skin friendly; dermatologically approved
Lightest weight ANSI A4/ISO Cut D glove featuring INTERCEPT™ Technology, these ultra-thin breathable gloves offer high cut resistance, while maintaining enhanced flexibility and tactility. Touchscreen compatible glove for enhanced productivity. Ultra-lightweightMore Product Information