Industrial gloves, EDGE® 48-128
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48-128/10 48-128/11 48-128/6 48-128/7 48-128/8 48-128/9
ANSE48-128/10EA 23.9 EUR
ANSE48-128/10 ANSE48-128/11 ANSE48-128/6 ANSE48-128/7 ANSE48-128/8 ANSE48-128/9
Industrial gloves, EDGE® 48-128
Handschoenen
Multipurpose, abrasion-resistant, nitrile-coated safety gloves offering oil resistance.
  • Very light and comfortable
  • High level of dexterity and flexibility
  • Good level of abrasion and oil resistance
  • Dark glove shows dirt contamination less quickly
  • FDA-compliant materials: Gloves are safe for food packaging
These flexible high-dexterity work gloves feature a lightweight, seamless knitted dark grey polyester liner and elasticized knitwrist dipped into black straight nitrile coating. Thanks to their nitrile coating, EDGE® 48-128 protective gloves grant better abrasion and oil resistance, grip and cushioning than light and mediumweight knitted cotton gloves.
Recommended for: Assembly of small parts; general handling; packaging and quality control.
Certificaten: EN 388:2016 (4121A); Cat. II