Cut resistant industrial gloves, EDGE® 48-702 series
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48-702/10 48-702/11 48-702/6 48-702/7 48-702/8 48-702/9
ANSE48-702/10EA 111 EUR
ANSE48-702/10 ANSE48-702/11 ANSE48-702/6 ANSE48-702/7 ANSE48-702/8 ANSE48-702/9
Cut resistant industrial gloves, EDGE® 48-702 series
Handschoenen
Besides providing excellent cut and abrasion protection and a firm grip, especially in dry and slightly oily environments, EDGE® 48-702 microfoam nitrile gloves also ensure higher tactile sensitivity and dexterity compared to foam nitrile gloves.
  • Seamless liner made from HPPE, Nylon, Polyester and Spandex
  • Coated palm is thinner than most comparable NBR gloves
  • Breathable coating keeps hands cool and dry
  • Composed of FDA-compliant materials, these safety gloves are suitable for handling and packaging food
Recommended for: Automotive, machinery and equipment, metal fabrication, oil and gas.
Certificaten: EN 388:2016 (4X42B); Cat. II