Oil resistant industrial gloves, EDGE® 48-919
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48-919/10 48-919/11 48-919/6 48-919/7 48-919/8 48-919/9
ANSE48-919/10EA 49.6 EUR
ANSE48-919/10 ANSE48-919/11 ANSE48-919/6 ANSE48-919/7 ANSE48-919/8 ANSE48-919/9
Oil resistant industrial gloves, EDGE® 48-919
Handschoenen
Reinforced with two layers of nitrile coating and a rough palm, EDGE® 48-919 light duty work gloves combine superior grip with abrasion and oil repellence and keep hands dry and comfortable when handling oily parts and tools.
  • Good combination of grip and oil repellence
  • Excellent abrasion resistance
  • Double layer nitrile coating keeps hands dry and comfortable when handling oily parts and tools
  • Elastic knit wrist ensures a snug fit
  • Gloves comply with FDA food handling requirements
Gloves feature a seamless liner and are fully NBR dipped to provide protection against industrial fluid exposure. The rough NBR palm coating ensures a highly secure grip in oily environment. Designed for workers who handle small and medium-sized components coated with oil or lubricant.
Certificaten: EN 388:2016 (4121A), Cat. II
Food contact approved