Industrial natural rubber latex gloves, AlphaTec® 87-085 series
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87-085/7 87-085/8 87-085/9 87-085/10
ANSE87-085/7EA 27.5 EUR
ANSE87-085/7 ANSE87-085/8 ANSE87-085/9 ANSE87-085/10
Industrial natural rubber latex gloves, AlphaTec® 87-085 series
Handschoenen
Lightweight natural rubber gloves offering comfort and food compliance.
  • Beaded cuff for tear resistance and easy donning
  • Food contact approved
  • Good abrasion resistance
  • 100% cotton flock lining
  • AQL 0.65
Flock lined, lightweight natural rubber latex gloves are designed for use in low hazard areas. Resistant, flexible, and very comfortable.