Trousers, AlphaTec® MICROGARD 2000
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WH20B-00301-02 WH20B-00301-04 WH20B-00301-05 WH20B-00301-06 WH20B-00301-03
ANSEWH20B-00301-02EA 350 EUR
ANSEWH20B-00301-02 ANSEWH20B-00301-04 ANSEWH20B-00301-05 ANSEWH20B-00301-06 ANSEWH20B-00301-03
Trousers, AlphaTec® MICROGARD 2000
Werkkleding
Superior breathable microporous laminate technology.
  • Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
  • Comfortable to wear, featuring moisture vapour permeable fabric to reduce the risk of heat stress
  • Optimized fit, improving wearer comfort and safety
  • Low-linting
The MICROGARD 2000 range is designed to allow water vapour (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals and filter 100% of particulates down to 0,01 microns in size. Bound seams offer superior strength, as well as a liquid and particle barrier.
Trousers feature elasticating to waist and ankles, and bound seams.