Industrial PVC gloves, EDGE® 14-663
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14-663/8 14-663/10 14-663/9
ANSE14-663/8EA 417 EUR
ANSE14-663/8 ANSE14-663/10 ANSE14-663/9
Industrial PVC gloves, EDGE® 14-663
Handschoenen
Fully-coated with PVC, EDGE® 14-663 safety gloves ensure greater flexibility and comfort while guarding hands against chemical, mechanical and biological hazards, making them ideal for workers exposed to oils, greases, acids, caustics and alcohols.
  • The EDGE range offers a greater flexibility, when compared to other models in its category
  • Highly cost-effective chemical protection PVC glove
  • Combines excellent protection against several chemical products with mechanical resistance
  • Non-slip finish on the palm, fingers, and back of hand for a better grip on wet
More Product Information
Order Now
Specificaties
Ambidexter/handspecifiek Handspecifiek
Environmentally Preferable
Voor eenmalig gebruik/herbruikbaar Herbruikbaar
Latex/latexvrij Latexvrij
Materiaal PVC
Afwerking Fully coated, rough finish
Manchetstijl Lang (Gauntlet)
Handpalmdikte 1,65 mm
EN-norm EN ISO 374-1:2016 Type C, EN ISO 374-5: 2016, EN 388:2016 (3121A)
Sterility Non sterile