Industrial gloves, EDGE® 48-701
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48-701/10 48-701/11 48-701/6 48-701/7 48-701/8 48-701/9
ANSE48-701/10EA 91.7 EUR
ANSE48-701/10 ANSE48-701/11 ANSE48-701/6 ANSE48-701/7 ANSE48-701/8 ANSE48-701/9
Industrial gloves, EDGE® 48-701
Handschoenen
EDGE® 48-701 industrial gloves' palm PU coating provides a high level of abrasion protection (ANSI 6), moderate cut resistance (ANSI A2), a good grip in dry environments, and better protection against oil than knitted or cut and sewn safety work gloves.
  • High cut protection and abrasion level
  • Good grip in dry environments
  • Palm dipping for good protection against oil
  • Seamless liner made from HPPE, Spandex, Polyester
  • Knitted wrist for form-fitting comfort, good dexterity and added wrist protection
EDGE® 48-701 gloves offer two levels of cut performance (depending on the application), a high level of abrasion resistance, and good sensitivity and fit. A cost-effective alternative compared to medium weight leather, thick cotton, or aramid gloves.
Recommended for: Aeronautics, assembly line, assembling metal parts and components, handling sharp or irregular metal pieces, molding, plastic injections.
Certificaten: EN 388:2016 (4342B), Cat. II