Industrial gloves, EDGE® 48-703 series
Leverancier: Ansell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
48-703/10 48-703/11 48-703/9 48-703/7 48-703/8
ANSE48-703/10EA 310 EUR
ANSE48-703/10 ANSE48-703/11 ANSE48-703/9 ANSE48-703/7 ANSE48-703/8
Industrial gloves, EDGE® 48-703 series
Handschoenen
Advanced cut protection combined with premium leather for superior grip.
  • Good dexterity and flexibility
  • Medium cut protection and high levels of abrasion resistance
  • Good grip in dry and slightly oily environments
  • Leather pad provides extra safety in grip, cut/abrasion protection, and cushioning during stamping applications
Gloves are made with a seamless liner and a knitted wrist to provide a close fit in cut resistant applications. The EDGE 48-703 series combines a good level of cut, abrasion, and thermal resistance in one glove. In addition, itMore Product Information
Order Now
Specificaties
Model
Ambidexter/handspecifiek Handspecifiek Handspecifiek
Environmentally Preferable
Voor eenmalig gebruik/herbruikbaar Herbruikbaar Herbruikbaar
Latex/latexvrij Latex Latex
Gepoederd/poedervrij poedervrij poedervrij
Voeringmateriaal Material2-21408841|Material2-21408168 Material2-21408841|Material2-21408168
Materiaal Leer Leer
Afwerking Leather palm
Steken per inch (Gauge) 13 13
Manchetstijl Gebreide manchet Gebreide manchet
Sterility Non sterile Non sterile