Disposable culture tubes, rimless, dispenser pack and bulk pack, PYREX®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
99445-10 99445-12 99445-13 99445-16 99445-16X 99445-16XX 99445-18 99445-20
734-4221EA 56.91 EUR
734-4221 734-4222 734-4223 734-4225 734-4226 734-4227 734-4228 734-4229
Disposable culture tubes, rimless, dispenser pack and bulk pack, PYREX®
Buisjes Cultuurbuisjes
Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.
These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed.