Part enclosed shelves
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MWF0550/0350 MWF0600/0400 MWF0600/0600
TEKMMWF0550/0350EA 0 EUR
TEKMMWF0550/0350 TEKMMWF0600/0400 TEKMMWF0600/0600
Part enclosed shelves
Meubilair Legborden
Our wall shelf is manufactured in a dull polish finish and available in various sizes.
  • Brushed dull polish finish
  • Wall fixed
Our stainless steel wall shelf with sides is manufactured from 304 grade stainless steel in a brushed dull polish finish. The enclosed design ensures that these are ideal for microwaves as well as other appliances that need to be stored safely.