Disposable flat bottom culture tubes, screw cap style, bulk pack, PYREX®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
99448-16X 99448-16 99448-19
734-4237EA 429.16 EUR
734-4237 734-4236 734-4238
Disposable flat bottom culture tubes, screw cap style, bulk pack, PYREX®
Buisjes Cultuurbuisjes
Tube has flat bottom and comes without marking spot. Tubes are made of borosilicate glass and are sold without caps.