Advanced MB, InduProof®
Leverancier: GETT GERATETECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KG17203 KG17204
GETTKG17203EA 327 EUR
GETTKG17203 GETTKG17204
Advanced MB, InduProof®
Toetsenborden
Rugged, water- and dust-proof silicone keyboard with integrated mouse button.
  • Rugged, durable keyboard with 104 keys and integrated mouse button
  • Comfortable & ergonomic operation also with protective gloves made of silicone or latex
  • IP 68 protection, can be submerged into liquids
  • Easy cleaning and disinfection - even under tap water or by immersion
  • Increased chemical and mechanical resistance due to special surface treatment
  • VESA-M5-boreholes (distance 75×75 mm)
  • 4 years warranty on technical function
  • Made in Germany
The InduProof® Advanced has a full set of keys with a numeric keyboard, an integrated mouse button (mini-joystick) and is a convincing product thanks to its pleasant sense of touch and the very high degree of protection that it provides. As a result of its robust design, it is highly suitable for use in challenging, hygiene-sensitive areas like industrial food and drinks production. It is possible to completely clean and disinfect the unit. It has a long serviceable life as a result of its high-quality workmanship – after all it is 'Made in Germany'.
Certificaten: CE, REACH, RoHs, EMC, IP 68.
Verpakking: The keyboards are packed in brown cardboard boxes.