Sink with drainer and undershelf
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SKDS-15X60L2 SKDS-15X65L SKDS-18X65L2
TEKMSKDS-15X60L2EA 0 EUR
TEKMSKDS-15X60L2 TEKMSKDS-15X65L TEKMSKDS-18X65L2
Sink with drainer and undershelf
Meubilair Omkasting
This stainless-steel sink with under shelf is available with left hand, right hand, or double drainer, to ensure that the product fits your requirement.
  • 304 grade stainless steel bowl with 430 stainless steel frame and shelf
  • Brushed dull polish finish
  • Crevice free, easily cleaned surfaces
  • Complete with waste, plug, chain, and overflow
  • Inset rear legs: 100 mm
  • Floor clearance: 237 mm
  • Upstand: 50 mm
  • 28 mm diameter taps holes with 180 mm centres
It comes in an easy to assemble, flat pack format. Its fully welded 304 grade stainless steel bowl and 430 frame and shelf, with its brushed dull polish finish, makes it ideal for industries where hygiene requirements must be at the forefront when buying furniture and equipment.
Furthermore, its stainless-steel adjustable feet provide stability to the sink. Although taps are not included, 28 mm diameter tap holes with 180 mm centres allow for convenient installation of taps and the sink unit comes complete with waste, plug, chain, and overflow.
The clever design and manufacturing of this stainless-steel sink has a crevice free surface for easy cleaning. Moreover, the undershelf will provide you with some extra storage space.