Spinner flasks with vertical sidearms, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4510-15L 4510-36L 4510-8L 4512-15L 4512-36L 4512-8L
734-1926EA 2296.2 EUR
734-1926 734-1927 734-1928 734-1929 734-1930 734-1931
Spinner flasks with vertical sidearms, Corning®
Kolven Spinnerflessen
Borosilicate glass flasks are baffled to enhance agitation of contents.
  • Side baffles enhance aeration and agitation of flask contents
  • Proprietary 'stretch' impeller design ensures optimal stirring
  • Vertical sidearms permit use with sidearm probes and fittings
Can be used with direct drive motor or magnetically driven paddle assembly.
Consistent height to width ratios for greater predictability in scale up across the full range of vessel sizes.