Floor cupboards with drawers
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SHCB-FD60X84 SHCB-FD12X84 SHCB-FD15X84 SHCB-FD18X84 SHCB-F31C12X84
TEKMSHCB-FD60X84EA 0 EUR
TEKMSHCB-FD60X84 TEKMSHCB-FD12X84 TEKMSHCB-FD15X84 TEKMSHCB-FD18X84 TEKMSHCB-F31C12X84
Floor cupboards with drawers
Meubilair Omkasting
Our floor cupboard comes with a drawer fitted as standard and in a range of sizes.
  • Brushed dull polish finish
  • Stainless steel adjustable feet with non-marking rubber base
  • Lockable cupboard section
  • Mobile version available
  • Hinged door
  • Drawer(s)
We offer a floor cupboard to suit your needs within our flexible range of stainless steel floor cupboards. Manufactured from 304-grade stainless steel our cupboards are both sturdy and practical.
The range of cupboards comes with an intelligent design to fit our standard range of tables (840 mm) for modularMore Product Information