Hanging racks for clipboards
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WW0215/6 WW0218/4 WW0218/6
TEKMWW0215/6EA 0 EUR
TEKMWW0215/6 TEKMWW0218/4 TEKMWW0218/6
Hanging racks for clipboards
Meubilair Omkleedrekken
Our hanging racks are designed for standard portrait clipboards.
  • Brushed dull polish finish
  • Wall fixed
  • Clipboards not included
  • Suitable for standard portrait clipboards with or without back clip
They come in two variations for compatibility with either standard portrait clipboards or portrait clipboards with back clips.
The hanging racks for clipboards are manufactured from fully welded 304 grade stainless steel in a brushed dull polish finish and are designed to be wall fixed.