CK4 wireless capacitive glass keyboards, Cleankeys®
Leverancier: GETT GERATETECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KR24230 KR24231 KR25207 KR25209 KR25205 KR25208 KR25206 KR25211 KR25210
GETTKR24230EA 519 EUR
GETTKR24230 GETTKR24231 GETTKR25207 GETTKR25209 GETTKR25205 GETTKR25208 GETTKR25206 GETTKR25211 GETTKR25210
CK4 wireless capacitive glass keyboards, Cleankeys®
Toetsenborden
Wireless capacitive glass keyboard with touchpad and number pad.
  • Fully enclosed Gorilla® glass surface
  • IP 65 rating (frontside)
  • 2,4 GHz wireless connection
  • Washable and disinfectable; therefore a high level of hygiene and cleanliness
  • Keylock function for cleaning the keyboard
  • Complete key layout with integrated touchpad
  • Volume and sensitivity individually adjustable
  • VESA mounting on the back
  • Four years warranty on the electronic function
  • Made in Germany
Cleankeys® is the brand for designer keyboards with a touch surface made of glass. They stand outMore Product Information