Mobile safety steps
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MST01000-3 MST01250-4 MST01500-5 MST0750-2
TEKMMST01000-3EA 0 EUR
TEKMMST01000-3 TEKMMST01250-4 TEKMMST01500-5 TEKMMST0750-2
Mobile safety steps
Ladders en opstapjes
Our mobile safety steps offer optimal durability and toughness, being fully welded in 304 grade stainless steel, offering a non-slip surface for maximum safety.
  • Whole structure comes with a brushed dull polish finish
  • Stainless steel non slip check plate steps to ensure maximum safety and security for workers
  • 3 castors- 2 swivel to rear and 1 retractable fixed at front
  • Locking handle
  • Widened legs
The steps castors and swivel ensure ease of manoeuvre, and the locking handle secures the steps ready for use. Different heights are available.