Sterile goods baskets
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PBK0600/300 PBK0600/150 PBK0600/300/A
TEKMPBK0600/300EA 0 EUR
TEKMPBK0600/300 TEKMPBK0600/150 TEKMPBK0600/300/A
Sterile goods baskets
Manden Draadmanden
High-quality electro polished 304 grade stainless steel baskets, with a sterile design for maximum hygiene.
  • Available with front access allows for easy access of stored goods
  • Stronger welds for extra weight capacity and rigidity
  • Wire structure allows optimal airflow
  • Modular system stackable to maximise space saving
Fully welded in 304 grade stainless steel for maximum durability and corrosion resistance, it has an electro polished finish for ease of cleaning and to prevent build-up of residue. Available with sterile goods divider.
Order Now
Specificaties
Environmentally Preferable