Enclosed Waste Bins
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HWWB-FSFE HWWB-FSFEFOL HWWB-FSFEFOLC HWWB-FSFEL HWWB-MBFEFOL
TEKMHWWB-FSFEEA 0 EUR
TEKMHWWB-FSFE TEKMHWWB-FSFEFOL TEKMHWWB-FSFEFOLC TEKMHWWB-FSFEL TEKMHWWB-MBFEFOL
Enclosed Waste Bins
Bakken Afvalbakken
The enclosed waste bins lid is supplied with tough elastic bands.
  • Brushed dull polish finish
  • Mobile version available
  • Strong and sturdy design
  • Supplied ready to use
  • Foot operated lids available for easy operation
Our enclosed waste bins are manufactured from 304-grade stainless steel. A plastic part free design and food grade edge trims ensure our products are easy to clean, supporting rigorous hygiene standards. A tough elastic band securely holds the bag in place, reducing the risk of tearing or spilling waste.
Mobile versions that offer even more useability are available whilst ensuring hygiene standards are maintained.
Bestelinformatie: Available without a lid or choose from our range of lift lids and foot operated, to aid with hands-free use, are available.