Floor cupboards
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SHCB-F60X84 SHCB-FM60X84 SHCB-F12X84 SHCB-FM12X84 SHCB-FU12X84 SHCB-F15X84 SHCB-FM15X84 SHCB-FU15X84 SHCB-FUM15X84 SHCB-F18X84 SHCB-FM18X84
TEKMSHCB-F60X84EA 0 EUR
TEKMSHCB-F60X84 TEKMSHCB-FM60X84 TEKMSHCB-F12X84 TEKMSHCB-FM12X84 TEKMSHCB-FU12X84 TEKMSHCB-F15X84 TEKMSHCB-FM15X84 TEKMSHCB-FU15X84 TEKMSHCB-FUM15X84 TEKMSHCB-F18X84 TEKMSHCB-FM18X84
Floor cupboards
Meubilair Opslagkasten
Our floor cupboards come in a range of sizes.
  • Brushed dull polish finish
  • Stainless steel adjustable feet with non-marking rubber base
  • Lockable cupboard
  • Mobile version available
  • Hinged door
The cupboards enable you to make full use of the roomy storage space inside, or you can create more surface storage by adding an optional shelf by dropping it onto the easy clean welded studs at 325 mm above the base/floor. These studs are positioned so the shelf will feature a very slight backward lean so to minimise the risk of water pooling and Listeria build up. The small gap between the shelf and the back of the cupboard then lets the water fall away via the drainage holes at the back of the base. We have thought of everything so that Listeria cannot lurk here!
Dirt ledges are minimised, by being easily swept away from the insides of the door(s) and the vertical frame on the back and sides offer no harbourage or dirt ledges for microbes to collect and multiply.
Our range of cupboards are intelligently designed to stand alongside our standard range of workbenches and tables at 840 mm high for perfect modular compatibility. They feature four non marking rubber base adjustable feet for perfect balance or you can go mobile for easy access. The anti-snag handle ensures you will keep your items safe (spare key included), and the option of upstand ensures items will not disappear at the back and an ideal storage solution for hygienic production areas.
To support safety and quick cleaning, consider adding a 20 mm deep drip tray at the bottom to catch any substances and stop them spreading around.