Open Frame Pedestal Waste Bins
Leverancier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WB0000 WB0005 WB0010 WB0010/CR
TEKMWB0000EA 0 EUR
TEKMWB0000 TEKMWB0005 TEKMWB0010 TEKMWB0010/CR
Open Frame Pedestal Waste Bins
Bakken Afvalbakken
Our hygienic and robust stainless steel open frame waste bag holder with a range of lids.
  • Mobile version available
  • Strong and sturdy design
  • Supplied with tough elastic bands or stainless steel clamping bands (cleanroom version)
  • Foot operated lids available for easy operation
Our open frame waste bag holders are manufactured from 304-grade stainless steel, ensuring optimised durability, corrosion resistance and hygiene.
A plastic free design and food grade edge trims ensure our products are easy to clean, supporting rigorous hygiene standards.
A tough elastic band securely holds the bag in place, reducing the risk of tearing or spilling waste. For the clean room versions this is replaced by a stainless steel clamping band.
Our open frame waste bag holders are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durable and easy to clean product ideal for your workspace.