Ethyl 3-(tert-butyl)-1-(2,4,6-trimethylbenzyl)-1H-pyrazole-5-carboxylate
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
OR2479-1G OR2479-5G
APOSOR2479-1GEA 96.4 EUR
APOSOR2479-1G APOSOR2479-5G
Ethyl 3-(tert-butyl)-1-(2,4,6-trimethylbenzyl)-1H-pyrazole-5-carboxylate
Ethyl 3-(tert-butyl)-1-(2,4,6-trimethylbenzyl)-1H-pyrazole-5-carboxylate
Formule: C₂₀H₂₈N₂O₂
Smeltpunt: 72…74 °C
MDL Nummer: MFCD01571363
CAS nummer: 306936-99-2