Polypropylene closures, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26795-25 26796-38 26797-38
CORN26795-25EA 114.26 EUR
CORN26795-25 CORN26796-38 CORN26797-38
Polypropylene closures, Corning®
Sluitingen Reageerbuissluitingen
These Corning® polypropylene closures are designed for culture tubes and flasks.
  • Internal drip ring minimizes contamination
  • Autoclavable at 121 °C
Closure offers two positions: open for gas exchange or closed for humidified environment.