Lightweight Plastic Pallets
Leverancier: MULTIROIR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
REP.1208.A REP.1210.A 1208PASGEN1 1210PAS
MULCREP.1208.AEA 44.2 EUR
MULCREP.1208.A MULCREP.1210.A MULC1208PASGEN1 MULC1210PAS
Lightweight Plastic Pallets
Dozen Transportdozen
Lightweight plastic pallets are made from recycled PP or HDPE and are a real alternative to wooden pallets.
  • Material: Recycled PP
  • Security lip 6 mm
  • Temperature from −20 to +40 °C
  • Open deck
These pallets are both lightweight and sturdy. They are available in sizes 120×80 and 120×100 with 4 feet or 3 skids. The 4 feet pallets are nestable and stackable.