Sealing mats for assay and storage microplates, Axygen®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AM-2ML-RD AM-2ML-RD-S AM-2ML-SQ AM-384-DW-SQ AM-500UL-RD AM-750UL-RD AM-24-SQ-S
736-0340EA 85.2 EUR
736-0340 731-0638 736-0346 736-0333 736-0337 732-1026 AXYGAM-24-SQ-S
Sealing mats for assay and storage microplates, Axygen®
Afdichtingen Afdichtingen van microtiterplaten
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications.
  • Minimize evaporation
  • Made of research-grade silicone
  • Autoclavable
They can be reused following a bleach wash and ethanol rinse protocol.