Union Fittings, Hose Barb, Tee, Polypropylene, Dynalon®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX06294-10BLKEA 81.7 EUR
MFLX06294-10BLK MFLX06294-30BLK MFLX06294-40BLK MFLX06294-60BLK
Union Fittings, Hose Barb, Tee, Polypropylene, Dynalon®
Fittingen en connectoren
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).