Union Fittings, Hose Barb, Y-Connector, Polyproylene, Dynalon®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX06295-10BLKEA 81.7 EUR
MFLX06295-10BLK MFLX06295-20BLK MFLX06295-30BLK MFLX06295-40BLK MFLX06295-50BLK MFLX06295-60BLK MFLX06295-70BLK MFLX06296-00BLK MFLX06296-20BLK MFLX06296-50BLK
Union Fittings, Hose Barb, Y-Connector, Polyproylene, Dynalon®
Fittingen en connectoren
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.