Bags, stand-up, recyclable, PE/EVOH-PE
Leverancier: Flexico
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ITM080501 ITM080502 ITM080503 ITM080504 ITM080505 ITM080506 ITM080507 ITM080508
129-2303EA 358 EUR
129-2303 129-2304 129-2305 129-2306 129-2307 129-2308 129-2309 129-2310
Bags, stand-up, recyclable, PE/EVOH-PE
Zakken Monsterzakken
Clear bags, zip seal, food contact.
  • Oxygen barrier and protection against humidity
  • Ideal for greasy or dry products
  • Suitable for food contact
  • Transparency and zip seal with sound and tactile feel
  • Resealable, reusable, 100% recyclable
  • Upright position for shelf display
The PE/EVOH-PE bag is a 100% recyclable mono-material complex film. The presence of a 5 µm EVOH layer provides excellent barrier protection against oxygen, carbon dioxide and retains aromas, without impacting the recyclability of the bag. The layers of polyethylene surrounding the EVOH also provide the bag with a humidity protection. Your products are protected from external contamination and their properties are preserved.
Certificaten: Suitable for food contact. Certificate available on request.