Bags, stand-up, PET/ALU/PE, Doypack®
Leverancier: Flexico
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ITM080203 ITM080204 ITM080207 ITM080201 ITM080202 ITM080205 ITM080206
129-2383EA 430 EUR
129-2383 129-2384 129-2387 129-2381 129-2382 129-2385 129-2386
Bags, stand-up, PET/ALU/PE, Doypack®
Zakken Monsterzakken
Bags, zip seal, food contact.
  • Optimum preservation of your products
  • Ideal for dry, wet or greasy content
  • Suitable for food contact
  • Zip seal with sound and tactile feel
  • Resealable and reusable
  • Upright position for shelf display
The PET/ALU/PE laminate gives to the bag an optimum preservation of your products. Aluminium is the best barrier against humidity, gases, UV and heat. It is a perfect solution for the conservation of flavours.
Certificaten: Suitable for food contact. Certificate available on request.