Stands for Transferpette® electronic Pipettes
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
705390 705391 705385 705386 705384 705383
613-4226EA 212 EUR
613-4226 613-4227 613-3720 613-3721 613-6937 613-6938
Stands for Transferpette® electronic Pipettes
Pipethouders
Save space in the laboratory and use stands to safely store your BRAND single or multichannel pipettes.
The single stand is ideal for storing one Transferpette® electronic pipette - one stand for pipettes up to 1000 µl and another for the 5 mL pipette.
Three Transferpette® electronic pipettes (single channel, up to 1000 μl) can be charged simultaneously in the charging stand. A separate power supply is no longer needed.