Keyboard for hygienic environments, InduProof® Smart Classic
Leverancier: GETT GERATETECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KG22293 KG24206 KG29000 KG29001
GETTKG22293EA 175 EUR
GETTKG22293 GETTKG24206 GETTKG29000 GETTKG29001
Keyboard for hygienic environments, InduProof® Smart Classic
Toetsenborden
Fully waterproof and dustproof medical keyboard with number pad.
  • Dustproof and waterproof keyboard
  • Can be completely disinfected
  • Soft typing through silicone switching elements
  • Long lifetime thanks to strict quality control and high quality materials
  • Ergonomic, space-saving design for maximum comfort during operation
  • Three year warranty on technical function
  • Made in Germany
The InduProof® Smart Classic has a full set of keys with a numeric keypad and offers a pleasant sense of touch and a very high degree of protection. The keyboard is ideal for use in almost any hygiene sensitive area. It can be completely cleaned and disinfected. A long service life is a result of its high quality workmanship.
Certificaten: CE, FCC, REACH, RoHs, EMC, IP 68.
Verpakking: Keyboards are safely and environmently friendly packed in brown cardboard boxes.