Silicone encased USB computer mouse for hygienic areas, GCQ® PrimeMouse
Leverancier: GETT GERATETECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KH29213 KH29214
GETTKH29213EA 64.8 EUR
GETTKH29213 GETTKH29214
Silicone encased USB computer mouse for hygienic areas, GCQ® PrimeMouse
Computermuizen en muismatten
The PrimeMouse offers ergonomic functionality with two mouse buttons and 3 scroll buttons. Due to its symmetrical housing shape, it is perfectly suitable for both right-handed and left-handed users. The PrimeMouse also has a completely waterproof and dustproof silicone housing.
  • Ergonomical functionality with two mouse buttons and three scroll buttons
  • Symmetrical shaped silicone housing is suited for right and left hand operation
  • IP 68 Silicone mouse with fully waterproof and dustproof housing
  • Optical sensor with a resolution of 1000 dpi
  • With plastic base plate, available in white or black
  • Easy to clean and to disinfect
  • Silicone cap to protect the USB plug against splash during cleaning
This silicone mouse has an ergonomic design and a pleasant tactile feeling with a high protection rating (IP 68). It can be easily cleaned and disinfected all around. The mouse is a product from the GCQ® series by GETT.
Certificaten: CE, FCC (class B, part 15), REACH, RoHs, EMC, IP 68. Medical-standard EMC certification: EN 60601-1-2 Edition 4. Industry-standard EMC certification: EN IEC 61000-6-4 (emission standard) and EN IEC 61000-6-2 (immunity).
Verpakking: The mouse is delivered safely and environmentally friendly packed in brown cardboard box.