Plastic keyboards for hygienic sensitive environments, GCQ CleanType® Easy Basic
Leverancier: GETT GERATETECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KL25205 KL25203 KL25206 KL25211 KL25212 KL26211 KL26212 KL26213 KL26215 KL26216
GETTKL25205EA 73.4 EUR
GETTKL25205 GETTKL25203 GETTKL25206 GETTKL25211 GETTKL25212 GETTKL26211 GETTKL26212 GETTKL26213 GETTKL26215 GETTKL26216
Plastic keyboards for hygienic sensitive environments, GCQ CleanType® Easy Basic
Toetsenborden
The CleanType® Easy Basic is a full layout, high-quality plastic keyboard that is easy to clean thanks to its water and dust resistance.
  • Washable plastic keyboard
  • Waterproof and dustproof
  • Available with white or black finish
  • 15 cm short cable with USB protection cap
  • Additional USB extension cable provided
  • 4 LED indicators (Num, scroll, caps lock, clean mode)
The CleanType® Easy Basic is a high-quality plastic keyboard that is easy to clean thanks to its water and dust resistance, making it ideal for use in hygiene-sensitive areas such as hospitals, clinics, medical practices, laboratories, or pharmaceutical companies. The CleanType® Easy Basic keyboard can be customized by having an individualized company logo lasered on it, integrating it perfectly into a current corporate identity. In addition, this keyboard can be equipped with any existing country layout.
Multiple cleaning methods are possible for this keyboard series. For dry cleaning, use a cleaning brush to remove dust. For wet cleaning, clean the keyboard under running warm water (max. 60 °C) with a cleaning brush (if required with commercially available cleaning agents).
Certificaten: CE, FCC, REACH, IP 68.
Bestelinformatie: Includes USB extension cable.
Verpakking: Keyboards are safely and environmentally friendly packed in brown cardboard boxes.