Plastic keyboards for hygienic sensitive environments, GCQ CleanType® Easy Basic Compact
Leverancier: GETT GERATETECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KL29203 KL29204 KL29205 KL29206
GETTKL29203EA 65 EUR
GETTKL29203 GETTKL29204 GETTKL29205 GETTKL29206
Plastic keyboards for hygienic sensitive environments, GCQ CleanType® Easy Basic Compact
Toetsenborden
The CleanType® Easy Basic Compact is a dust-protected compact plastic keyboard, for use in confined spaces without sacrificing important functions.
  • Available with white or black finish
  • 15 cm short cable with USB protection cap
  • USB extension cable (+174 cm) included
  • LED indicators for scroll, shift, and key lock
  • Keylock function can be switched on and off by pressing the 'Clean on/off' button
The CleanType® Easy Basic Compact is a high-quality compact plastic keyboard that is easy and thorough to clean, making it ideal for use in hygiene sensitive areas such as hospitals, clinics, doctors, offices, laboratories, or pharmaceutical companies.
Upon request, the CleanType® Easy Basic Compact can be equipped with any desired country layout, even in small quantities. In addition, it can be laser-engraved with a company logo. This service provides an individual product that can be perfectly integrated into an existing corporate identity.
Certificaten: CE, FCC, REACH.
Bestelinformatie: Includes USB extension cable.
Verpakking: Keyboards are delivered safely and environmentally friendly packed in brown cardboard.