Static dissipative facemasks
Leverancier: DESCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
97550 97551 97552 97553 97554 97555
DESC97550EA 22.2 EUR
DESC97550 DESC97551 DESC97552 DESC97553 DESC97554 DESC97555
Static dissipative facemasks
Maskers Hygiëne maskers
These facemasks constructed of a lightweight dissipative material which is made from texturised polyester and a minimum of 9% carbon nylon monofiliment.
  • Can be washed multiple times in a mild detergent
  • Elastic ear loops
  • Adjustable nose wire helps ensure mask fits securely
  • Acceptable for use in ESD protected areas
  • Provides continuous and consistent charge dissipation
Elastic bands range from small-medium (69,85 mm) to large-extra large (82,55 mm). The mask area is the same size for all items.