Metal-In Shielding Bubble Bags
Leverancier: DESCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
200515 200530 200540
129-2615EA 7 EUR
129-2615 129-2616 129-2617
Metal-In Shielding Bubble Bags
Zakken Opbergzakken
  • Combines 'Faraday Cage' and mechanical protection
  • Shields about twice as well as normal shielding bag of equivalent size
  • Flap can be tucked in to enclose contents or folded down and held in place by a label
  • Inner and outer surface resistance: 1×104 to <1×1011 ohms; T₁₀₀₀: <2 sec at 50% rH