Anti-static Pink Poly Bags with Write-On Panel
Leverancier: DESCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
203000 203015
129-2627EA 14.4 EUR
129-2627 129-2628
Anti-static Pink Poly Bags with Write-On Panel
Zakken Monsterzakken
For use with non-ESD sensitive items destined for use in an EPA.
  • Economical
  • Made of polyethylene
  • RS: 1×10⁵ to <1×10¹¹ ohms, T₁₀₀₀: <2 s at 50% rH, 22 °C