Smiths Medical® Luer Fittings on PVC Flexible Tubing, Straight
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX30526-14BLKEA 204 EUR
MFLX30526-14BLK MFLX30526-16BLK
Smiths Medical® Luer Fittings on PVC Flexible Tubing, Straight
Fittingen en connectoren
Get a seamless transition from source to system.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
This high pressure flexible PVC tubing has luer fittings at each end and comes in various lengths. Can be sterilized by ETO.