Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MFLX30800-08BLKEA 407 EUR
MFLX30800-08BLK MFLX45508-00BLK MFLX30800-06BLK
Value Plastics® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene
Fittingen en connectoren
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.