Depth stop for samplers
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5317-0015 5317-0020
BURK5317-0015EA 169 EUR
BURK5317-0015 BURK5317-0020
Depth stop for samplers
Monsternemers Monsternemers voor vaste stoffen
To make a sampling procedure reproducible, it should always be carried out under the same conditions. For this purpose, the depth stop for samplers is a helpful accessory.
  • Enables standardised and thus reproducible sampling
  • Very good clamping effect due to C-clamping screw
  • Available for Ø 25/50mm, suitable for many sampler lance
The depth stop can be clamped to any height of the sampler by means of a C-clamping screw. This ensures that the sampler penetrates to the same desired depth each time it is pierced. The penetration depth is thus fixed and canMore Product Information