Gaussia-firefly luciferase dual assay kit, Pierce™, Thermo Scientific
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16181 16182
PIER16181EA 167 EUR
PIER16181 PIER16182
Gaussia-firefly luciferase dual assay kit, Pierce™, Thermo Scientific
Testen Enzyme-assays
The Thermo Scientific Pierce Gaussia-Firefly Luciferase dual assay kit provides the necessary reagents to simultaneously detect intracellular Gaussia and red firefly luciferase activity in mammalian whole cell lysates.
  • Simultaneous—concurrent, filter-based, wavelength-separated detection of two luciferase activities
  • Sensitive—measure bright blue Gaussia and red firefly luciferase activities in the same sample
  • Fast—no quenching step required, unlike in traditional sequential dual assays
  • Multiplex—capable of quantitating two cellular activities in the same sample(s)
Gaussia luciferase acts as an experimental reporter with constitutively active red firefly as a normalisation control. The assay working solution contains substrates for both luciferases, and the reactions occur simultaneously with flash-type kinetics. The resulting luminescent signals are spectrally resolvable using filters. In a single sample, researchers can assay transcriptional activity of regulatory elements, signal transduction pathways, and effects of activators or inhibitors.
Leveringsinformatie: Kit includes Gaussia-Firefly dual assay buffer (5 ml), 1X Coelenterazine (5 µl), D-Luciferin, Lyophilised (3 mg) and 2X Cell Lysis Buffer (6 ml).
Opgelet: For research use only. Not for use in diagnostic procedures.