Erlenmeyer Flasks with Screw Neck
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
81003016 81004049 81004050
LENZ81003016EA 58.8 EUR
LENZ81003016 LENZ81004049 LENZ81004050
Erlenmeyer Flasks with Screw Neck
Kolven Erlenmeyers
DURAN®, amber coloured, with DIN thread.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to rapid temperature changes
With white volume graduations and white marking field. Amber coloured to protect light-sensitive substances.