Fragment Analyzer Parts and Accessories
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P60-20 P60-22
AGLSP60-20EA 402 EUR
AGLSP60-20 AGLSP60-22
Fragment Analyzer Parts and Accessories
Elektroforesesystemen Systemen voor capillaire elektroforese
Search for genuine spare parts and accessories for the Fragment Analyzer systems to keep your operations running. Find out more here.
  • 96-well deep well tray - for buffer and waste plates
  • Tray carrier adaptor - molded plate adaptor designed to hold semi-skirted PCR plate in the drawer location
  • Tube cap and bottle cap replacement - modified for liquid tubing
  • High-quality, genuine Agilent consumables ensure low instrument downtime and best total cost of ownership
Obtain genuine spare parts and accessories specifically designed to ensure continued high level of instrument performance for the Agilent Fragment Analyzer systems.
The portfolio includes tray carrier, tube cap, gel bottle cap replacement and 96 deep-well trays. In addition, a tray adaptor for customers wanting to use 12 vial strip tubes is available.
Opgelet: For research use only. Not for use in diagnostic procedures.