ZAG dsDNA Kits
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ZAG-105-5000 ZAG-110-5000 ZAG-130-5000 ZAG-135-5000
AGLSZAG-105-5000EA 2350 EUR
AGLSZAG-105-5000 AGLSZAG-110-5000 AGLSZAG-130-5000 AGLSZAG-135-5000
ZAG dsDNA Kits
Elektroforesesystemen Systemen voor capillaire elektroforese
High-throughput PCR fragment sizing and microsatellite detection.
  • Broad concentration range - All kits have a concentration range of 0,5 to 50 ng/µl
  • Reliable sizing - Four different kits to choose from with a sizing range varying from 1 to 20000 bp
  • Low sample volume - Only 2 µl of sample is required, minimising precious sample loss to QC steps
  • Quick separations - Run times as short as 30 minutes
  • High resolution - Separate PCR fragments under 300 bp with a 3 bp difference or greater
The ZAG dsDNA kits provide a high-throughput analysis solution for relative quantification and accurate sizing of dsDNA fragments. Specially formulated gels, markers, and ladders are designed for each kit enabling reliable sizing over a broad range with varying applications. Common applications include PCR fragment sizing, genotyping, and microsatellite detection.
There are four ZAG dsDNA kits to choose from based on the size of the DNA or PCR fragment of interest, ranging from 1 to 20000 bp. Results can be obtained in as little as 30 minutes while maintaining high resolution.
Opgelet: For research use only. Not for use in diagnostic procedures.