Cell-free DNA ScreenTape Analysis (ScreenTape and Reagents)
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5067-5630 5067-5631 5067-5632 5067-5633
AGLS5067-5630EA 358 EUR
AGLS5067-5630 AGLS5067-5631 AGLS5067-5632 AGLS5067-5633
Cell-free DNA ScreenTape Analysis (ScreenTape and Reagents)
Elektroforesesystemen Systemen voor capillaire elektroforese
Integrity assessment of cell-free DNA samples, with the detection of high molecular weight DNA contaminations.
  • Quantification and sizing of cell-free DNA from 50 to 800 bp and detect High Molecular Weight (HMW) DNA contamination
  • cfDNA sample detection is possible down to 20 pg/µl
  • Digital assessment of %cfDNA to qualify sample integrity
  • Results may be obtained in less than 2 minutes per sample
  • Input sample QC allows saving of sequencing and sample preparation in Next Generation Sequencing workflows
  • Ready to use ScreenTape devices and automated data analysis provide an easy-to-use system with excellent reproducibility
  • Partially used ScreenTape devices can be used for completion at a later point in time
Cell-free DNA ScreenTape analysis provides fast, automated and reliable electrophoretic separation of input cell-free DNA (cfDNA) samples. The Cell-free DNA ScreenTape assay provides accurate quantification and sizing as well as the value %cfDNA.
The %cfDNA is an objective and reproducible value for the assessment of sample quality. It automatically calculates the amount of cfDNA in the sample of interest. Delivering digital assessment of cfDNA concentration, size and quality in under two minutes per sample.
The assay is compatible with the Agilent 4150 and 4200 TapeStation instruments.
Opgelet: For research use only. Not for use in diagnostic procedures.