Bioanalyzer Parts and Accessories
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5065-4401 5065-4413 5065-9951
AGLS5065-4401EA 1260 EUR
AGLS5065-4401 AGLS5065-4413 AGLS5065-9951
Bioanalyzer Parts and Accessories
Elektroforesesystemen Systemen voor capillaire elektroforese
Bioanalyzer spare parts, including electrode cartridges, priming stations, filters, and fuses.
  • Removable electrode cartridges allow labs to buy additional cartridges dedicated for specific applications
  • Replacement test chip kits are available for troubleshooting and running diagnostic tests
  • The chips are reusable and expire after three years
  • Multiple chip priming stations are available and may be useful for core facilities or labs with multiple Bioanalyser users
  • To ensure proper maintenance of the Bioanalyzer system additional gaskets are available in the gasket kit
  • Cables and connectors for the Bioanalyser system may also be bought separately
Maximise the efficiency of your Bioanalyzer system with genuine Agilent accessories and spare parts. Our extensive portfolio includes electrode cartridges and priming stations for high use labs running many Bioanalyzer assays.
In addition, if you're running short on parts we stock extra test chips, electrode cleaners, gasket adapters, filters, cables and fuses. Our genuine Agilent accessories are precision-engineered and available separately to ensure your applications remain versatile, uninterrupted and safe.
Opgelet: For research use only. Not for use in diagnostic procedures.