TapeStation Systems Parts and Accessories
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5067-5598 5067-5599 5067-5783 5067-5786 5188-8047 G2991-40007 G2992-40042 G2992-40046
732-3993EA 43.9 EUR
732-3993 732-3994 AGLS5067-5783 AGLS5067-5786 AGLS5188-8047 AGLSG2991-40007 AGLSG2992-40042 AGLSG2992-40046
TapeStation Systems Parts and Accessories
Elektroforesesystemen Systemen voor capillaire elektroforese
Accessories and spare parts for the 4150 and 4200 TapeStation systems like plates & foil seals, loading tips, TapeStation test tape, needle cartridge.
  • 8-well PCR tube strips or 96-well sample plates are available depending on sample throughput, bringing added flexibility
  • Needle cartridges enable fast and simple needle exchange to ensure proper maintenance of the TapeStation systems
  • TapeStation test tape is available for troubleshooting and running system diagnostics tests, the tapes are reusable and expire after two years
  • Loading tips are pre-packed and ready to use
Our portfolio of TapeStation systems spare parts and accessories features everything needed to keep your lab productive. Maximise the efficiency of your TapeStation systems today with our genuine agilent accessories.
The portfolio features additional needle cartridges and TapeStation test tape. In addition, if running short on spare parts, extra loading tips are also available, along with 96-well sample plates and foil seals. For enhanced flexibility we also offer 8-well PCR optical tube strips and caps.
Opgelet: For research use only. Not for use in diagnostic procedures.